Assay Design



Gene Transcripts

Gene Symbol
TGM5
Entrez Gene ID
9333
Gene Name
transglutaminase 5
Gene Aliases
PSS2, TGASE5, TGASEX, TGM6, TGMX, TGX
Chromosome Location
Chr.15: 43232595 - 43266857 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201631.3 NP_963925.2 2 - 3 203 82
GenBank mRNA AF035960.1 - 2 - 3 82
BC119009.1 - 2 - 3 203 82
BC122859.1 - 2 - 3 203 82

View Full Product Details