Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004245.3 | NP_004236.1 | 4 - 5 | 444 | 101 |
NM_201631.3 | NP_963925.2 | 5 - 6 | 690 | 101 | |
GenBank mRNA | AF035960.1 | - | 5 - 6 | 690 | 101 |
AF035961.1 | - | 4 - 5 | 444 | 101 | |
AK298479.1 | - | 1 - 2 | 106 | 101 | |
BC119009.1 | - | 5 - 6 | 690 | 101 | |
BC122859.1 | - | 5 - 6 | 690 | 101 |