Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004245.3 | NP_004236.1 | 6 - 7 | 756 | 115 |
NM_201631.3 | NP_963925.2 | 7 - 8 | 1002 | 115 | |
GenBank mRNA | AF035960.1 | - | 7 - 8 | 1002 | 115 |
AF035961.1 | - | 6 - 7 | 756 | 115 | |
AK298479.1 | - | 3 - 4 | 418 | 115 | |
BC119009.1 | - | 7 - 8 | 1002 | 115 | |
BC122859.1 | - | 7 - 8 | 1002 | 115 |