Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256552.1 | NP_001243481.1 | 1 - 2 | 406 | 107 |
XM_006720678.3 | XP_006720741.1 | 1 - 2 | 357 | 107 | |
XM_006720680.1 | XP_006720743.1 | 2 - 3 | 497 | 107 | |
XM_011522007.1 | XP_011520309.1 | 2 - 3 | 497 | 107 | |
XM_017022568.1 | XP_016878057.1 | 1 - 2 | 364 | 107 | |
GenBank mRNA | BC010876.1 | - | 1 - 2 | 387 | 107 |