Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198713.1 | NP_001185642.1 | 7 - 8 | 1013 | 98 |
NM_001256782.1 | NP_001243711.1 | 8 - 9 | 1110 | 98 | |
NM_001256822.1 | NP_001243751.1 | 7 - 8 | 1082 | 98 | |
NM_004574.4 | NP_004565.1 | 7 - 8 | 1213 | 98 | |
NM_080415.3 | NP_536340.1 | 6 - 7 | 1097 | 98 | |
NM_080416.3 | NP_536341.1 | 7 - 8 | 984 | 98 | |
NR_037155.2 | 6 - 7 | 1581 | 98 | ||
NR_104196.1 | 7 - 8 | 1213 | 98 | ||
NR_104197.1 | 6 - 7 | 1107 | 98 | ||
XM_006721949.2 | XP_006722012.1 | 8 - 9 | 1042 | 98 | |
XM_006721950.3 | XP_006722013.1 | 7 - 8 | 734 | 98 | |
XM_006721951.2 | XP_006722014.1 | 6 - 7 | 2868 | 98 | |
XM_006721952.2 | XP_006722015.1 | 6 - 7 | 687 | 98 | |
XM_006721954.2 | XP_006722017.1 | 7 - 8 | 727 | 98 | |
XM_006721955.2 | XP_006722018.1 | 7 - 8 | 2910 | 98 | |
XM_011524911.1 | XP_011523213.1 | 6 - 7 | 641 | 98 | |
XM_011524912.1 | XP_011523214.1 | 7 - 8 | 1399 | 98 | |
GenBank mRNA | AB002110.1 | - | 6 - 7 | 1384 | 98 |
AB008753.1 | - | 7 - 8 | 1048 | 98 | |
AF035811.1 | - | 7 - 8 | 1049 | 98 | |
AF073312.1 | - | 7 - 8 | 1044 | 98 | |
AF176379.1 | - | 6 - 7 | 1063 | 98 | |
AK094579.1 | - | 6 - 7 | 2105 | 98 | |
AK294094.1 | - | 7 - 8 | 1016 | 98 | |
AK301914.1 | - | 8 - 9 | 1110 | 98 | |
AK302146.1 | - | 7 - 8 | 1013 | 98 | |
AK315045.1 | - | 7 - 8 | 98 | ||
AK315396.1 | - | 7 - 8 | 938 | 98 | |
BC018055.1 | - | 6 - 7 | 1024 | 98 | |
BC018056.1 | - | 7 - 8 | 927 | 98 | |
CR457111.1 | - | 7 - 8 | 920 | 98 | |
D89278.1 | - | 7 - 8 | 920 | 98 | |
U88829.1 | - | 7 - 8 | 1045 | 98 | |
U88870.1 | - | 7 - 8 | 930 | 98 |