Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198713.1 | NP_001185642.1 | 10 - 11 | 1366 | 54 |
NM_001256782.1 | NP_001243711.1 | 11 - 12 | 1463 | 54 | |
NM_001256822.1 | NP_001243751.1 | 10 - 11 | 1435 | 54 | |
NM_004574.4 | NP_004565.1 | 10 - 11 | 1566 | 54 | |
NM_080416.3 | NP_536341.1 | 10 - 11 | 1337 | 54 | |
NR_104197.1 | 9 - 10 | 1460 | 54 | ||
XM_006721949.2 | XP_006722012.1 | 11 - 12 | 1395 | 54 | |
XM_006721950.3 | XP_006722013.1 | 10 - 11 | 1087 | 54 | |
XM_006721951.2 | XP_006722014.1 | 9 - 10 | 3221 | 54 | |
XM_006721952.2 | XP_006722015.1 | 9 - 10 | 1040 | 54 | |
XM_006721954.2 | XP_006722017.1 | 10 - 11 | 1080 | 54 | |
XM_006721955.2 | XP_006722018.1 | 10 - 11 | 3263 | 54 | |
XM_011524911.1 | XP_011523213.1 | 9 - 10 | 994 | 54 | |
XM_011524912.1 | XP_011523214.1 | 10 - 11 | 1752 | 54 | |
GenBank mRNA | AB008753.1 | - | 10 - 11 | 1401 | 54 |
AF035811.1 | - | 10 - 11 | 1402 | 54 | |
AF073312.1 | - | 10 - 11 | 1397 | 54 | |
AK094579.1 | - | 9 - 10 | 2458 | 54 | |
AK294094.1 | - | 10 - 11 | 1369 | 54 | |
AK301914.1 | - | 11 - 12 | 1463 | 54 | |
AK302146.1 | - | 10 - 11 | 1366 | 54 | |
AK315396.1 | - | 10 - 11 | 1291 | 54 | |
BC018056.1 | - | 10 - 11 | 1280 | 54 | |
CR457111.1 | - | 10 - 11 | 1273 | 54 | |
D89278.1 | - | 10 - 11 | 1273 | 54 | |
U88829.1 | - | 10 - 11 | 1398 | 54 | |
U88870.1 | - | 10 - 11 | 1283 | 54 |