Assay Design



Gene Transcripts

Gene Symbol
SEPT4
Entrez Gene ID
5414
Gene Name
septin 4
Gene Aliases
ARTS, BRADEION, CE5B3, H5, MART, PNUTL2, SEP4, hCDCREL-2, hucep-7
Chromosome Location
Chr.17: 58520250 - 58540818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006721949.2 XP_006722012.1 2 - 3 188 102

View Full Product Details