Assay Design



Gene Transcripts

Gene Symbol
MLST8
Entrez Gene ID
64223
Gene Name
MTOR associated protein, LST8 homolog
Gene Aliases
GBL, GbetaL, LST8, POP3, WAT1
Chromosome Location
Chr.16: 2205177 - 2209417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199174.1 NP_001186103.1 1 - 2 147 150
GenBank mRNA AK304390.1 - 1 - 2 427 148

View Full Product Details