Assay Design



Gene Transcripts

Gene Symbol
ROGDI
Entrez Gene ID
79641
Gene Name
rogdi homolog
Gene Aliases
KTZS
Chromosome Location
Chr.16: 4796962 - 4802950 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024589.2 NP_078865.1 2 - 3 502 57
NR_046480.1 2 - 3 502 57
XM_006720947.3 XP_006721010.1 2 - 3 183 57
XM_006720948.3 XP_006721011.1 2 - 3 185 57
GenBank mRNA AK026039.1 - 2 - 3 225 57
AK098348.1 - 2 - 3 185 57
AL136675.1 - 2 - 3 209 57
BC012901.2 - 2 - 3 170 57
CR457355.1 - 2 - 3 123 57
CR533547.1 - 2 - 3 123 57

View Full Product Details