Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000697.2 | NP_000688.2 | 2 - 3 | 391 | 88 |
XM_011523780.2 | XP_011522082.1 | 2 - 3 | 2665 | 88 | |
GenBank mRNA | BC069557.1 | - | 2 - 3 | 369 | 88 |
M35418.1 | - | 2 - 3 | 379 | 88 | |
M58704.1 | - | 2 - 3 | 371 | 88 | |
M62982.1 | - | 2 - 3 | 391 | 88 |