Assay Design



Gene Transcripts

Gene Symbol
EVPL
Entrez Gene ID
2125
Gene Name
envoplakin
Gene Aliases
EVPK
Chromosome Location
Chr.17: 76006845 - 76027452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320747.1 NP_001307676.1 15 - 16 2212 60
NM_001988.3 NP_001979.2 15 - 16 2146 60
GenBank mRNA AK290333.1 - 15 - 16 2146 60
BC126103.1 - 15 - 16 1925 60
BC126105.1 - 15 - 16 1925 60
BC143814.1 - 15 - 16 1991 60
U53786.1 - 15 - 16 1990 60

View Full Product Details