Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304284.1 | NP_001291213.1 | 22 - 23 | 3727 | 58 |
NM_004505.3 | NP_004496.2 | 14 - 15 | 3196 | 58 | |
XM_011524050.1 | XP_011522352.1 | 18 - 19 | 3945 | 58 | |
XM_011524051.2 | XP_011522353.1 | 20 - 21 | 3040 | 58 | |
XM_011524052.2 | XP_011522354.1 | 19 - 20 | 3154 | 58 | |
XM_011524053.2 | XP_011522355.1 | 20 - 21 | 3067 | 58 | |
XM_011524054.2 | XP_011522356.1 | 20 - 21 | 3034 | 58 | |
XM_011524055.2 | XP_011522357.1 | 18 - 19 | 2961 | 58 | |
XM_011524056.2 | XP_011522358.1 | 15 - 16 | 2914 | 58 | |
XM_011524057.2 | XP_011522359.1 | 18 - 19 | 3077 | 58 | |
XM_011524058.2 | XP_011522360.1 | 19 - 20 | 3143 | 58 | |
XM_011524059.2 | XP_011522361.1 | 19 - 20 | 3143 | 58 | |
XM_017025290.1 | XP_016880779.1 | 17 - 18 | 3783 | 58 | |
GenBank mRNA | AB449915.1 | - | 13 - 14 | 1497 | 58 |
AY143550.1 | - | 14 - 15 | 3196 | 58 | |
BX647719.1 | - | 22 - 23 | 3728 | 58 | |
X63546.1 | - | 14 - 15 | 3193 | 58 | |
X63547.1 | - | 15 - 16 | 3404 | 58 |