Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 24 - 25 | 3606 | 111 |
GenBank mRNA | AB209901.1 | - | 24 - 25 | 3591 | 111 |
AF035121.1 | - | 24 - 25 | 3607 | 111 | |
AF063658.1 | - | 24 - 25 | 3304 | 111 | |
BC131822.1 | - | 24 - 25 | 3366 | 111 | |
L04947.1 | - | 24 - 25 | 3298 | 111 | |
X61656.1 | - | 24 - 25 | 3298 | 111 |