Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 2 - 3 | 462 | 98 |
GenBank mRNA | AB209901.1 | - | 2 - 3 | 447 | 98 |
AF035121.1 | - | 2 - 3 | 463 | 98 | |
AF063658.1 | - | 2 - 3 | 160 | 98 | |
BC131822.1 | - | 2 - 3 | 222 | 98 | |
BP280621.1 | - | 2 - 3 | 462 | 98 | |
EU826563.1 | - | 2 - 3 | 160 | 98 | |
FJ899739.1 | - | 2 - 3 | 160 | 98 | |
L04947.1 | - | 2 - 3 | 154 | 98 | |
X61656.1 | - | 2 - 3 | 154 | 98 |