Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 | 5 - 6 | 950 | 110 |
GenBank mRNA | AB209901.1 | - | 5 - 6 | 935 | 110 |
AF035121.1 | - | 5 - 6 | 951 | 110 | |
AF063658.1 | - | 5 - 6 | 648 | 110 | |
BC131822.1 | - | 5 - 6 | 710 | 110 | |
EU826563.1 | - | 5 - 6 | 648 | 110 | |
FJ899739.1 | - | 5 - 6 | 648 | 110 | |
L04947.1 | - | 5 - 6 | 642 | 110 | |
X61656.1 | - | 5 - 6 | 642 | 110 |