Assay Design
Gene Transcripts
- Gene Symbol
- MOB2
- Entrez Gene ID
-
81532
- Gene Name
- MOB kinase activator 2
- Gene Aliases
-
HCCA2
- Chromosome Location
-
Chr.11: 1469448 - 1486779 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK057350.1 |
- |
3 - 4 |
437 |
67 |
View Full Product Details