Assay Design
Gene Transcripts
- Gene Symbol
- MOB2
- Entrez Gene ID
-
81532
- Gene Name
- MOB kinase activator 2
- Gene Aliases
-
HCCA2
- Chromosome Location
-
Chr.11: 1469448 - 1486779 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB066564.1 |
- |
2 - 3 |
334 |
63 |
AK300235.1 |
- |
1 - 2 |
192 |
63 |
View Full Product Details