Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024956.3 | NP_079232.3 | 2 - 3 | 570 | 107 |
XM_005254682.4 | XP_005254739.1 | 2 - 3 | 583 | 107 | |
XM_011522074.2 | XP_011520376.1 | 2 - 3 | 586 | 107 | |
XM_017022600.1 | XP_016878089.1 | 2 - 3 | 615 | 107 | |
XM_017022601.1 | XP_016878090.1 | 2 - 3 | 368 | 107 | |
XM_017022604.1 | XP_016878093.1 | 2 - 3 | 584 | 107 | |
XM_017022606.1 | XP_016878095.1 | 2 - 3 | 583 | 107 | |
XM_017022609.1 | XP_016878098.1 | 2 - 3 | 584 | 107 | |
XM_017022610.1 | XP_016878099.1 | 2 - 3 | 344 | 107 | |
XM_017022611.1 | XP_016878100.1 | 2 - 3 | 399 | 107 | |
GenBank mRNA | AK309679.1 | - | 1 - 2 | 86 | 107 |
BC009981.2 | - | 2 - 3 | 570 | 107 |