Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024956.3 | NP_079232.3 | 5 - 6 | 893 | 94 |
XM_005254682.4 | XP_005254739.1 | 4 - 5 | 860 | 94 | |
XM_005254683.4 | XP_005254740.1 | 3 - 4 | 749 | 94 | |
XM_011522071.1 | XP_011520373.1 | 4 - 5 | 377 | 94 | |
XM_011522072.2 | XP_011520374.1 | 3 - 4 | 544 | 94 | |
XM_017022600.1 | XP_016878089.1 | 4 - 5 | 892 | 94 | |
XM_017022601.1 | XP_016878090.1 | 4 - 5 | 645 | 94 | |
XM_017022602.1 | XP_016878091.1 | 3 - 4 | 781 | 94 | |
XM_017022603.1 | XP_016878092.1 | 4 - 5 | 377 | 94 | |
XM_017022604.1 | XP_016878093.1 | 4 - 5 | 861 | 94 | |
XM_017022605.1 | XP_016878094.1 | 3 - 4 | 750 | 94 | |
GenBank mRNA | AK027028.1 | - | 2 - 3 | 198 | 94 |
AK225353.1 | - | 2 - 3 | 198 | 94 | |
AK309679.1 | - | 4 - 5 | 409 | 94 | |
BC009981.2 | - | 5 - 6 | 893 | 94 |