Assay Design



Gene Transcripts

Gene Symbol
TMEM62
Entrez Gene ID
80021
Gene Name
transmembrane protein 62
Gene Aliases
-
Chromosome Location
Chr.15: 43133115 - 43185147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024956.3 NP_079232.3 8 - 9 1307 118
XM_005254682.4 XP_005254739.1 7 - 8 1274 118
XM_005254683.4 XP_005254740.1 6 - 7 1163 118
XM_005254684.3 XP_005254741.1 5 - 6 1020 118
XM_011522071.1 XP_011520373.1 7 - 8 791 118
XM_011522072.2 XP_011520374.1 6 - 7 958 118
XM_011522073.1 XP_011520375.1 4 - 5 456 118
XM_011522074.2 XP_011520376.1 7 - 8 1102 118
XM_017022600.1 XP_016878089.1 7 - 8 1306 118
XM_017022601.1 XP_016878090.1 7 - 8 1059 118
XM_017022602.1 XP_016878091.1 6 - 7 1195 118
XM_017022603.1 XP_016878092.1 7 - 8 791 118
XM_017022604.1 XP_016878093.1 7 - 8 1275 118
XM_017022605.1 XP_016878094.1 6 - 7 1164 118
XM_017022606.1 XP_016878095.1 6 - 7 1132 118
XM_017022607.1 XP_016878096.1 5 - 6 1052 118
XM_017022608.1 XP_016878097.1 5 - 6 586 118
XM_017022609.1 XP_016878098.1 6 - 7 1133 118
XM_017022610.1 XP_016878099.1 6 - 7 893 118
XM_017022611.1 XP_016878100.1 6 - 7 948 118
XM_017022612.1 XP_016878101.1 6 - 7 1067 118
XM_017022613.1 XP_016878102.1 5 - 6 759 118
XM_017022614.1 XP_016878103.1 5 - 6 717 118
XM_017022615.1 XP_016878104.1 5 - 6 698 118
XM_017022616.1 XP_016878105.1 5 - 6 756 118
GenBank mRNA AK027028.1 - 5 - 6 612 118
AK225353.1 - 5 - 6 612 118
AK309679.1 - 7 - 8 823 118
BC009981.2 - 8 - 9 1307 118
CR457370.1 - 3 - 4 404 118

View Full Product Details