Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098509.1 | NP_001091979.1 | 12 - 13 | 1616 | 69 |
NM_014853.2 | NP_055668.2 | 13 - 14 | 1753 | 69 | |
XM_011524101.2 | XP_011522403.1 | 14 - 15 | 1829 | 69 | |
XM_011524102.2 | XP_011522404.1 | 14 - 15 | 1827 | 69 | |
XM_011524103.2 | XP_011522405.1 | 14 - 15 | 1828 | 69 | |
XM_011524104.2 | XP_011522406.1 | 14 - 15 | 2353 | 69 | |
XM_011524105.2 | XP_011522407.1 | 13 - 14 | 1693 | 69 | |
XM_011524106.1 | XP_011522408.1 | 13 - 14 | 1651 | 69 | |
XM_011524107.2 | XP_011522409.1 | 14 - 15 | 1830 | 69 | |
XM_011524108.1 | XP_011522410.1 | 12 - 13 | 1516 | 69 | |
XM_017025474.1 | XP_016880963.1 | 13 - 14 | 1691 | 69 | |
XM_017025475.1 | XP_016880964.1 | 13 - 14 | 1692 | 69 | |
XM_017025476.1 | XP_016880965.1 | 8 - 9 | 988 | 69 | |
GenBank mRNA | AB007857.3 | - | 12 - 13 | 1616 | 69 |
AB275762.1 | - | 13 - 14 | 1588 | 69 | |
AB449878.1 | - | 12 - 13 | 1441 | 69 | |
AK124883.1 | - | 3 - 4 | 1493 | 69 | |
AK125203.1 | - | 2 - 3 | 645 | 69 | |
AK294955.1 | - | 12 - 13 | 1508 | 69 | |
BC039204.1 | - | 13 - 14 | 1753 | 69 |