Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018460.3 | NP_060930.3 | 8 - 9 | 857 | 122 |
XM_011511479.2 | XP_011509781.1 | 9 - 10 | 889 | 122 | |
XM_011511481.2 | XP_011509783.1 | 8 - 9 | 792 | 122 | |
XM_011511482.2 | XP_011509784.1 | 8 - 9 | 792 | 122 | |
XM_011511483.1 | XP_011509785.1 | 4 - 5 | 487 | 122 | |
XM_011511484.1 | XP_011509786.1 | 8 - 9 | 792 | 122 | |
XM_017004499.1 | XP_016859988.1 | 8 - 9 | 818 | 122 | |
XM_017004500.1 | XP_016859989.1 | 7 - 8 | 684 | 122 | |
XM_017004501.1 | XP_016859990.1 | 8 - 9 | 792 | 122 | |
GenBank mRNA | AF217522.1 | - | 8 - 9 | 123 | |
AK226014.1 | - | 8 - 9 | 792 | 122 | |
AY219338.1 | - | 8 - 9 | 795 | 122 | |
BC016701.2 | - | 8 - 9 | 763 | 122 | |
BC038976.1 | - | 8 - 9 | 762 | 122 |