Assay Design



Gene Transcripts

Gene Symbol
CASC1
Entrez Gene ID
55259
Gene Name
cancer susceptibility candidate 1
Gene Aliases
LAS1, PPP1R54
Chromosome Location
Chr.12: 25107057 - 25195177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319978.1 NP_001306907.1 2 - 3 175 112
XM_017019566.1 XP_016875055.1 3 - 4 344 112

View Full Product Details