Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021090.3 | NP_066576.1 | 8 - 9 | 877 | 77 |
NM_153050.2 | NP_694690.1 | 8 - 9 | 877 | 77 | |
NM_153051.2 | NP_694691.1 | 8 - 9 | 877 | 77 | |
XM_005261803.2 | XP_005261860.1 | 9 - 10 | 995 | 77 | |
XM_005261804.3 | XP_005261861.1 | 8 - 9 | 912 | 77 | |
XM_005261805.2 | XP_005261862.1 | 9 - 10 | 995 | 77 | |
XM_005261806.2 | XP_005261863.1 | 8 - 9 | 877 | 77 | |
XM_005261808.2 | XP_005261865.1 | 9 - 10 | 995 | 77 | |
XM_017029028.1 | XP_016884517.1 | 7 - 8 | 769 | 77 | |
XM_017029029.1 | XP_016884518.1 | 7 - 8 | 759 | 77 | |
XM_017029030.1 | XP_016884519.1 | 7 - 8 | 775 | 77 | |
XM_017029031.1 | XP_016884520.1 | 7 - 8 | 763 | 77 | |
XM_017029032.1 | XP_016884521.1 | 8 - 9 | 760 | 77 | |
XM_017029033.1 | XP_016884522.1 | 2 - 3 | 196 | 77 | |
XM_017029034.1 | XP_016884523.1 | 8 - 9 | 850 | 77 | |
GenBank mRNA | AB002369.1 | - | 8 - 9 | 796 | 77 |
AF233436.2 | - | 5 - 6 | 558 | 77 | |
AF233437.1 | - | 5 - 6 | 558 | 77 | |
AF233438.1 | - | 5 - 6 | 558 | 77 | |
AK289581.1 | - | 8 - 9 | 877 | 77 | |
BC142713.1 | - | 8 - 9 | 833 | 77 | |
BC148216.1 | - | 7 - 8 | 696 | 77 | |
BC151217.1 | - | 8 - 9 | 891 | 77 | |
BC152455.1 | - | 8 - 9 | 796 | 77 | |
CR456525.1 | - | 6 - 7 | 573 | 77 |