Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007245.2 | NP_001007246.1 | 9 - 10 | 1367 | 81 |
NM_001197079.1 | NP_001184008.1 | 8 - 9 | 1257 | 81 | |
NM_001197080.1 | NP_001184009.1 | 8 - 9 | 1092 | 81 | |
NM_001550.3 | NP_001541.2 | 8 - 9 | 1158 | 81 | |
NR_120333.1 | 7 - 8 | 920 | 81 | ||
XM_017012151.1 | XP_016867640.1 | 8 - 9 | 1405 | 81 | |
GenBank mRNA | AK297509.1 | - | 7 - 8 | 745 | 81 |
AK298894.1 | - | 8 - 9 | 898 | 81 | |
AK308066.1 | - | 9 - 10 | 1342 | 81 | |
AK315967.1 | - | 8 - 9 | 1090 | 81 | |
BC001272.1 | - | 8 - 9 | 1111 | 81 | |
BT019889.1 | - | 8 - 9 | 897 | 81 | |
BT019890.1 | - | 8 - 9 | 897 | 81 |