Assay Design
Gene Transcripts
- Gene Symbol
- PEX5L
- Entrez Gene ID
-
51555
- Gene Name
- peroxisomal biogenesis factor 5 like
- Gene Aliases
-
PEX5R, PEX5RP, PXR2, PXR2B, TRIP8b
- Chromosome Location
-
Chr.3: 179794958 - 180037053 on Build GRCh38
View Full Product Details