Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256750.1 | NP_001243679.1 | 2 - 3 | 541 | 80 |
XM_011512885.2 | XP_011511187.1 | 3 - 4 | 570 | 80 | |
XM_011512887.2 | XP_011511189.1 | 2 - 3 | 133 | 80 | |
GenBank mRNA | AB032592.1 | - | 2 - 3 | 258 | 80 |