Assay Design



Gene Transcripts

Gene Symbol
PEX5L
Entrez Gene ID
51555
Gene Name
peroxisomal biogenesis factor 5 like
Gene Aliases
PEX5R, PEX5RP, PXR2, PXR2B, TRIP8b
Chromosome Location
Chr.3: 179794958 - 180037053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256750.1 NP_001243679.1 7 - 8 1169 64
NM_001256751.1 NP_001243680.1 6 - 7 1103 64
NM_001256752.1 NP_001243681.1 6 - 7 1070 64
NM_001256753.1 NP_001243682.1 5 - 6 998 64
NM_001256754.1 NP_001243683.1 7 - 8 905 64
NM_001256755.1 NP_001243684.1 6 - 7 710 64
NM_001256756.1 NP_001243685.1 6 - 7 800 64
NM_016559.2 NP_057643.1 7 - 8 1175 64
XM_011512882.2 XP_011511184.1 8 - 9 1787 64
XM_011512884.2 XP_011511186.1 8 - 9 1783 64
XM_011512885.2 XP_011511187.1 8 - 9 1198 64
XM_011512886.2 XP_011511188.1 7 - 8 1004 64
XM_011512887.2 XP_011511189.1 6 - 7 566 64
XM_011512888.2 XP_011511190.1 6 - 7 866 64
XM_011512891.2 XP_011511193.1 5 - 6 1363 64
XM_011512892.2 XP_011511194.1 4 - 5 1169 64
XM_017006601.1 XP_016862090.1 7 - 8 1595 64
XM_017006602.1 XP_016862091.1 7 - 8 1000 64
XM_017006603.1 XP_016862092.1 7 - 8 1591 64
XM_017006604.1 XP_016862093.1 5 - 6 2181 64
XM_017006605.1 XP_016862094.1 5 - 6 866 64
XM_017006606.1 XP_016862095.1 7 - 8 1672 64
XM_017006607.1 XP_016862096.1 6 - 7 2369 64
XM_017006608.1 XP_016862097.1 6 - 7 1019 64
XM_017006609.1 XP_016862098.1 6 - 7 1512 64
XM_017006610.1 XP_016862099.1 5 - 6 947 64
XM_017006611.1 XP_016862100.1 5 - 6 1324 64
GenBank mRNA AB032592.1 - 7 - 8 886 64
AB032593.1 - 7 - 8 851 64
AK294227.1 - 4 - 5 400 64
AK294501.1 - 5 - 6 873 64
AK295349.1 - 6 - 7 568 64
AK295386.1 - 6 - 7 800 64
AK299633.1 - 6 - 7 976 64
AK303716.1 - 7 - 8 896 64
AK314283.1 - 7 - 8 1053 64
BC036183.1 - 6 - 7 954 64

View Full Product Details