Assay Design
Gene Transcripts
- Gene Symbol
- CRTAC1
- Entrez Gene ID
-
55118
- Gene Name
- cartilage acidic protein 1
- Gene Aliases
-
ASPIC, ASPIC1, CEP-68
- Chromosome Location
-
Chr.10: 97854711 - 98030828 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001182.1 |
- |
10 - 11 |
1316 |
102 |
View Full Product Details