Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031935.2 | NP_114141.2 | 70 - 71 | 11081 | 77 |
XM_011510038.2 | XP_011508340.1 | 70 - 71 | 11098 | 77 | |
XM_017002437.1 | XP_016857926.1 | 59 - 60 | 9031 | 77 | |
GenBank mRNA | AF156100.1 | - | 70 - 71 | 11084 | 77 |
AJ306906.1 | - | 13 - 14 | 1966 | 77 |