Assay Design



Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr.1: 185734526 - 186190953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031935.2 NP_114141.2 75 - 76 11793 140
XM_011510038.2 XP_011508340.1 75 - 76 11810 140
XM_017002437.1 XP_016857926.1 64 - 65 9743 140
GenBank mRNA AF156100.1 - 75 - 76 11796 140
AJ306906.1 - 18 - 19 2678 140

View Full Product Details