Assay Design



Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr.1: 185734526 - 186190953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031935.2 NP_114141.2 94 - 95 14988 103
XM_011510038.2 XP_011508340.1 94 - 95 15005 103
XM_017002437.1 XP_016857926.1 83 - 84 12938 103
GenBank mRNA AF156100.1 - 94 - 95 14991 103
AJ306906.1 - 37 - 38 5873 103
AK027344.1 - 4 - 5 547 103
AL833232.1 - 12 - 13 1937 103

View Full Product Details