Assay Design



Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr.1: 185734526 - 186190953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031935.2 NP_114141.2 105 - 106 16643 90
XM_011510038.2 XP_011508340.1 104 - 105 16309 90
XM_017002437.1 XP_016857926.1 94 - 95 14593 90
GenBank mRNA AF156100.1 - 105 - 106 16646 90
AJ306906.1 - 48 - 49 7528 90
AK027344.1 - 14 - 15 1851 90
AK096452.1 - 5 - 6 836 90
AL833232.1 - 22 - 23 3241 90

View Full Product Details