Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030787.3 | NP_110414.1 | 4 - 5 | 731 | 62 |
XM_011510020.2 | XP_011508322.1 | 4 - 5 | 638 | 62 | |
GenBank mRNA | AF295327.1 | - | 4 - 5 | 696 | 62 |
AK314026.1 | - | 4 - 5 | 732 | 62 | |
BC026282.1 | - | 4 - 5 | 712 | 62 | |
BC111773.1 | - | 4 - 5 | 696 | 62 |