Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030787.3 | NP_110414.1 | 5 - 6 | 913 | 80 |
XM_011510020.2 | XP_011508322.1 | 5 - 6 | 820 | 80 | |
GenBank mRNA | AF295327.1 | - | 5 - 6 | 878 | 80 |
AK314026.1 | - | 5 - 6 | 914 | 80 | |
BC026282.1 | - | 5 - 6 | 894 | 80 | |
BC111773.1 | - | 5 - 6 | 878 | 80 |