Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030787.3 | NP_110414.1 | 6 - 7 | 1099 | 133 |
XM_011510020.2 | XP_011508322.1 | 6 - 7 | 1006 | 133 | |
GenBank mRNA | AF295327.1 | - | 6 - 7 | 1064 | 133 |
AK314026.1 | - | 6 - 7 | 1100 | 133 | |
BC026282.1 | - | 6 - 7 | 1080 | 133 | |
BC111773.1 | - | 6 - 7 | 1064 | 133 |