Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000299.3 | NP_000290.2 | 11 - 12 | 2146 | 77 |
NM_001005337.2 | NP_001005337.1 | 10 - 11 | 2083 | 77 | |
GenBank mRNA | AK299473.1 | - | 2 - 3 | 218 | 77 |
AK315291.1 | - | 10 - 11 | 1904 | 77 | |
AK316131.1 | - | 2 - 3 | 218 | 77 | |
BC114571.1 | - | 10 - 11 | 1966 | 77 | |
BC115702.1 | - | 10 - 11 | 1728 | 77 | |
X79293.1 | - | 10 - 11 | 2020 | 77 | |
Z34974.1 | - | 10 - 11 | 2084 | 77 |