Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000299.3 | NP_000290.2 | 2 - 3 | 557 | 54 |
NM_001005337.2 | NP_001005337.1 | 2 - 3 | 557 | 54 | |
GenBank mRNA | AK315291.1 | - | 2 - 3 | 503 | 54 |
BC114571.1 | - | 2 - 3 | 440 | 54 | |
BC115702.1 | - | 2 - 3 | 440 | 54 | |
X79293.1 | - | 2 - 3 | 497 | 54 | |
Z34974.1 | - | 2 - 3 | 558 | 54 |