Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 3 - 4 | 594 | 72 |
NM_014456.4 | NP_055271.2 | 3 - 4 | 636 | 72 | |
NM_145341.3 | NP_663314.1 | 4 - 5 | 720 | 72 | |
GenBank mRNA | AK223623.1 | - | 3 - 4 | 510 | 72 |
AK296753.1 | - | 3 - 4 | 461 | 72 | |
AK312540.1 | - | 3 - 4 | 532 | 72 | |
AK315295.1 | - | 4 - 5 | 598 | 72 | |
BC015036.2 | - | 3 - 4 | 500 | 72 | |
BC026104.2 | - | 3 - 4 | 503 | 72 | |
BC031049.1 | - | 3 - 4 | 503 | 72 | |
BI910668.1 | - | 3 - 4 | 598 | 72 | |
BX537500.1 | - | 2 - 3 | 3584 | 72 | |
U96628.1 | - | 3 - 4 | 434 | 72 |