Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199492.1 | NP_001186421.1 | 6 - 7 | 1016 | 136 |
NM_014456.4 | NP_055271.2 | 6 - 7 | 1058 | 136 | |
NM_145341.3 | NP_663314.1 | 7 - 8 | 1142 | 136 | |
GenBank mRNA | AK223623.1 | - | 6 - 7 | 932 | 136 |
AK296753.1 | - | 6 - 7 | 883 | 136 | |
AK312540.1 | - | 6 - 7 | 954 | 136 | |
AK315295.1 | - | 7 - 8 | 1020 | 136 | |
BC026104.2 | - | 6 - 7 | 925 | 136 | |
BC031049.1 | - | 6 - 7 | 925 | 136 | |
BX641168.1 | - | 2 - 3 | 2451 | 136 | |
BX647834.1 | - | 6 - 7 | 900 | 136 | |
U96628.1 | - | 6 - 7 | 856 | 136 |