Assay Design



Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
Chromosome Location
Chr.1: 169511954 - 169586632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000130.4 NP_000121.2 1 - 2 305 148
GenBank mRNA AK226079.1 - 1 - 2 223 148
AK291613.1 - 1 - 2 255 148
AU279657.1 - 1 - 2 305 148
M14335.1 - 1 - 2 236 148
M16967.1 - 1 - 2 250 148

View Full Product Details