Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000130.4 | NP_000121.2 | 6 - 7 | 1099 | 132 |
XM_017000660.1 | XP_016856149.1 | 5 - 6 | 1108 | 132 | |
GenBank mRNA | AK226079.1 | - | 6 - 7 | 1017 | 132 |
AK291613.1 | - | 6 - 7 | 1049 | 132 | |
AK300469.1 | - | 5 - 6 | 957 | 132 | |
M14335.1 | - | 6 - 7 | 1030 | 132 | |
M16967.1 | - | 6 - 7 | 1044 | 132 |