Assay Design



Gene Transcripts

Gene Symbol
F5
Entrez Gene ID
2153
Gene Name
coagulation factor V
Gene Aliases
FVL, PCCF, RPRGL1, THPH2
Chromosome Location
Chr.1: 169511954 - 169586632 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000130.4 NP_000121.2 8 - 9 1435 138
XM_017000660.1 XP_016856149.1 7 - 8 1444 138
GenBank mRNA AK226079.1 - 8 - 9 1353 138
AK291613.1 - 8 - 9 1385 138
AK300469.1 - 7 - 8 1293 138
M14335.1 - 8 - 9 1366 138
M16967.1 - 8 - 9 1380 138

View Full Product Details