Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000130.4 | NP_000121.2 | 9 - 10 | 1542 | 88 |
XM_017000660.1 | XP_016856149.1 | 8 - 9 | 1551 | 88 | |
GenBank mRNA | AK226079.1 | - | 9 - 10 | 1460 | 88 |
AK291613.1 | - | 9 - 10 | 1492 | 88 | |
M14335.1 | - | 9 - 10 | 1473 | 88 | |
M16967.1 | - | 9 - 10 | 1487 | 88 |