Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199173.1 | NP_001186102.1 | 1 - 2 | 333 | 156 |
XM_005255479.2 | XP_005255536.1 | 1 - 2 | 253 | 156 | |
GenBank mRNA | AK098762.1 | - | 1 - 2 | 268 | 156 |
AY223837.1 | - | 1 - 2 | 154 | ||
DA382215.1 | - | 1 - 2 | 337 | 156 |