Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000590.1 | NP_000581.1 | 1 - 2 | 125 | 64 |
GenBank mRNA | BC066283.1 | - | 1 - 2 | 125 | 64 |
BC066284.1 | - | 1 - 2 | 125 | 64 | |
BC066285.1 | - | 1 - 2 | 125 | 64 | |
BC066286.1 | - | 1 - 2 | 125 | 64 | |
BC066287.1 | - | 1 - 2 | 125 | 64 | |
M30134.1 | - | 1 - 2 | 125 | 64 | |
S63356.1 | - | 1 - 2 | 125 | 64 | |
X17543.1 | - | 1 - 2 | 125 | 64 |