Assay Design



Gene Transcripts

Gene Symbol
GPR143
Entrez Gene ID
4935
Gene Name
G protein-coupled receptor 143
Gene Aliases
NYS6, OA1
Chromosome Location
Chr.X: 9725413 - 9765965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000273.2 NP_000264.2 8 - 9 1271 134
GenBank mRNA BC068977.1 - 8 - 9 1271 134
Z48804.1 - 8 - 9 1183 134

View Full Product Details