Assay Design



Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr.3: 46436005 - 46485234 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199149.1 NP_001186078.1 12 - 13 1694 109
NM_001321121.1 NP_001308050.1 12 - 13 1799 109
NM_001321122.1 NP_001308051.1 15 - 16 2038 109
NM_002343.5 NP_002334.2 12 - 13 1805 109
GenBank mRNA AF332168.1 - 12 - 13 1512 109
AK093852.1 - 15 - 16 2041 109
AK290859.1 - 12 - 13 1550 109
AK292813.1 - 12 - 13 1550 109
AK298035.1 - 12 - 13 1694 109
AK303995.1 - 4 - 5 413 109
AK316330.1 - 12 - 13 1614 109
AY137470.1 - 12 - 13 1512 109
AY165046.1 - 12 - 13 1524 109
AY178998.2 - 12 - 13 1512 109
AY360320.1 - 12 - 13 1507 109
AY493417.1 - 12 - 13 1622 109
AY875691.1 - 12 - 13 1512 109
BC015822.1 - 12 - 13 1554 109
BC015823.1 - 12 - 13 1556 109
BC022347.1 - 12 - 13 1550 109
EU779935.1 - 12 - 13 1512 109
EU794669.1 - 12 - 13 1547 109
KF992208.1 - 12 - 13 1524 109
KT006756.1 - 12 - 13 1512 109
M18642.1 - 2 - 3 207 109
M83202.1 - 12 - 13 1519 109
M83205.1 - 12 - 13 1498 109
M93150.1 - 12 - 13 1529 109
U07643.1 - 12 - 13 1609 109
X52941.1 - 12 - 13 1506 109
X53961.1 - 12 - 13 1806 109

View Full Product Details