Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000917.3 | NP_000908.2 | 13 - 14 | 1682 | 73 |
NM_001017962.2 | NP_001017962.1 | 13 - 14 | 1682 | 73 | |
NM_001142595.1 | NP_001136067.1 | 14 - 15 | 1775 | 73 | |
NM_001142596.1 | NP_001136068.1 | 12 - 13 | 1628 | 73 | |
GenBank mRNA | AK222960.1 | - | 13 - 14 | 1570 | 73 |
AK312593.1 | - | 13 - 14 | 1568 | 73 | |
BC034998.1 | - | 13 - 14 | 1575 | 73 | |
CD013929.1 | - | 12 - 13 | 1507 | 73 | |
CD013930.1 | - | 14 - 15 | 1654 | 73 | |
M24486.1 | - | 13 - 14 | 1559 | 73 | |
M24487.1 | - | 13 - 14 | 1559 | 73 |