Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181672.2 | NP_858058.1 | 13 - 14 | 1999 | 59 |
NM_181673.2 | NP_858059.1 | 13 - 14 | 1969 | 59 | |
XM_017029907.1 | XP_016885396.1 | 11 - 12 | 1528 | 59 | |
XM_017029908.1 | XP_016885397.1 | 11 - 12 | 5001 | 59 | |
GenBank mRNA | AF223393.1 | - | 9 - 10 | 1636 | 59 |
AK313530.1 | - | 9 - 10 | 1915 | 59 | |
AL050366.1 | - | 8 - 9 | 3226 | 59 | |
AL833085.2 | - | 13 - 14 | 1979 | 59 | |
BC014434.1 | - | 13 - 14 | 1900 | 59 | |
BC038180.1 | - | 13 - 14 | 1932 | 59 | |
BX537844.1 | - | 13 - 14 | 1957 | 59 | |
U77413.1 | - | 9 - 10 | 1648 | 59 |